siRNA Details
Gene Symbol: Rwdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147146.2 | NP_671487.1 | 2 | 259 | ||
XM_008772869.1 | XP_008771091.1 | 2 | 391 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025614, AK005278, AK003262, AK009510, AK011277, and others.
