siRNA Details
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148889.2 | NP_683687.1 | 5 | 595 | ||
XM_006238026.2 | XP_006238088.1 | 5 | 951 | |||
XM_006238027.2 | XP_006238089.1 | 4 | 987 | |||
XM_006238028.2 | XP_006238090.1 | 4 | 834 | |||
XM_006238029.2 | XP_006238091.1 | 3 | 168 | |||
XM_008763607.1 | XP_008761829.1 | 3 | 846 | |||
GenBank mRNA | AF398235.1 | - | 4 | 341 | ||
BC078716.1 | - | 5 | 595 | 7132044 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025444, NM_025545, NM_001025445, BC071213, AK005286, and others.
Gene Ontology Categories:
