siRNA Details
Gene Symbol: Drgx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145767.1 | NP_665710.1 | 4, 5 | 520 | ||
GenBank mRNA | U29174.1 | - | 4, 5 | 520 |

More Information
Other Species Targeted

Gene Ontology Categories:
