siRNA Details
Gene Symbol: HERC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016323.3 | NP_057407.2 | 9, 10 | 1399 | ||
XM_011532022.2 | XP_011530324.2 | 9, 10 | 1912 | |||
GenBank mRNA | AB027289.1 | - | 9, 10 | 1359 | ||
AK302142.1 | - | 3, 4 | 271 | |||
BC140716.1 | - | 9, 10 | 1292 | 9021584 |
