siRNA Details
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148889.2 | NP_683687.1 | 7 | 1003 | ||
XM_006238026.2 | XP_006238088.1 | 7 | 1359 | |||
XM_006238027.2 | XP_006238089.1 | 6 | 1395 | |||
XM_006238028.2 | XP_006238090.1 | 6 | 1242 | |||
XM_006238029.2 | XP_006238091.1 | 5 | 576 | |||
XM_008763607.1 | XP_008761829.1 | 5 | 1254 | |||
GenBank mRNA | AF398235.1 | - | 6 | 749 | ||
BC078716.1 | - | 7 | 1003 | 7132044 | ||
FQ214915.1 | - | 3 | 376 |
