siRNA Details
Gene Symbol: Ly49i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152848.1 | NP_690061.1 | 2 | 282 | ||
GenBank mRNA | AY115572.1 | - | 2 | 282 |
Gene Symbol: Ly49i5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009501.2 | NP_001009501.1 | 234 | |||
GenBank mRNA | AY649836.1 | - | 234 | |||
AY653729.1 | - | 79 | ||||
AY846264.1 | - | 84 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
