siRNA Details
Gene Symbol: Ly49s3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153726.1 | NP_714948.1 | 7 | 712 | ||
GenBank mRNA | AY102036.1 | - | 7 | 712 | ||
AY747628.1 | - | 8 | 763 |
Gene Symbol: Ly49i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009499.1 | NP_001009499.1 | 584 | |||
GenBank mRNA | AY659932.1 | - | 584 |
Gene Symbol: Ly49i4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009495.1 | NP_001009495.1 | 5 | 671 | ||
XM_008763405.1 | XP_008761627.1 | 6 | 786 | |||
GenBank mRNA | AY649834.1 | - | 5 | 671 |
Gene Symbol: Ly49s4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009487.1 | NP_001009487.1 | 712 | |||
GenBank mRNA | AY649831.1 | - | 712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
