siRNA Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153821.1 | NP_722543.1 | 1 | 423 | ||
XM_006250151.2 | XP_006250213.1 | 1 | 1161 | |||
GenBank mRNA | AY730764.1 | - | 1 | 147 | ||
S82911.1 | - | 1 | 423 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011127, NM_001025570, NM_175686, L06502, AB221686, and others.
Gene Ontology Categories:
