siRNA Details
Gene Symbol: Brs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152845.1 | NP_690058.1 | 1 | 255 | ||
XM_006257677.2 | XP_006257739.1 | 2 | 285 | |||
XM_006257678.2 | XP_006257740.1 | 2 | 104 | |||
GenBank mRNA | AF510984.1 | - | 1 | 255 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
