siRNA Details
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153622.2 | NP_705886.1 | 4 | 489 | ||
XM_006253941.2 | XP_006254003.1 | 7 | 1142 | |||
XM_006253942.2 | XP_006254004.1 | 7 | 1144 | |||
XM_006253944.2 | XP_006254006.1 | 7 | 1131 | |||
XM_006253946.2 | XP_006254008.1 | 6 | 1018 | |||
XM_006253947.2 | XP_006254009.1 | 7 | 1148 | |||
XM_006253948.2 | XP_006254010.1 | 2 | 1019 | |||
XM_006253949.2 | XP_006254011.1 | 5 | 901 | |||
XM_008771623.1 | XP_008769845.1 | 8 | 1219 | |||
XM_008771624.1 | XP_008769846.1 | 8 | 1207 | |||
XM_008771625.1 | XP_008769847.1 | 6 | 977 | |||
GenBank mRNA | AB025223.1 | - | 4 | 513 | ||
AB025224.1 | - | 4 | 513 | |||
BC078710.1 | - | 4 | 407 | 7110091 |
