siRNA Details
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153724.2 | NP_714946.2 | 2 | 307 | ||
XM_006248041.2 | XP_006248103.1 | 2 | 507 | |||
GenBank mRNA | AB075973.1 | - | 2 | 263 | ||
BC072478.1 | - | 2 | 307 | 7103498 |
