siRNA Details
Gene Symbol: Vom1r90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153729.2 | NP_714951.1 | 3 | 949 | ||
GenBank mRNA | AY510354.1 | - | 1 | 721 | ||
U36898.1 | - | 3 | 969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
