siRNA Details
Gene Symbol: Cd247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205304.1 | NP_001192233.1 | 4, 5 | 367 | ||
NM_170789.2 | NP_740770.2 | 5, 6 | 448 | |||
XM_006250147.2 | XP_006250209.1 | 5, 6 | 488 | |||
GenBank mRNA | BC097933.1 | - | 5, 6 | 391 | 7384006 | |
CO562119.1 | - | 4, 5 | 301 | |||
D13555.1 | - | 5, 6 | 448 | |||
L08447.1 | - | 5, 6 | 375 |
