siRNA Details
Gene Symbol: HOXC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017410.2 | NP_059106.2 | 2 | 930 | ||
GenBank mRNA | AF255676.1 | - | 2 | 930 | ||
AF263466.2 | - | 2 | 887 | |||
AK024027.1 | - | 2 | 412 | |||
BC002754.1 | - | 2 | 877 | |||
BC090850.1 | - | 2 | 899 | 6171228 | ||
BT009908.1 | - | 2 | 815 | |||
BU158600.1 | - | 2 | 219 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010464, AF193796, AY349616.
Gene Ontology Categories:
