siRNA Details
Gene Symbol: TAOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004783.3 | NP_004774.1 | 19 | 4234 | ||
XM_011545984.1 | XP_011544286.1 | 19 | 4481 | |||
XM_011545985.1 | XP_011544287.1 | 19 | 4478 | |||
GenBank mRNA | AB020688.1 | - | 19 | 3820 | ||
AK057589.1 | - | 2 | 1849 | |||
BC152413.1 | - | 19 | 3820 | 8862518 | ||
CN286840.1 | - | 1 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
