siRNA Details
Gene Symbol: Mrgprx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172158.2 | NP_750845.2 | 1 | 177 |
Gene Symbol: Mrgprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002286.1 | NP_001002286.1 | 1 | 135 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
