siRNA Details
Gene Symbol: Zfp455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173314.2 | NP_775436.2 | 2 | 165 | ||
XM_006232101.2 | XP_006232163.1 | 2 | 401 | |||
GenBank mRNA | BC161833.1 | - | 2 | 165 | ||
U67083.1 | - | 153 |

More Information
Other Species Targeted

Gene Ontology Categories:
