siRNA Details
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176074.3 | NP_788263.1 | 2, 3 | 310 | ||
XM_008760747.1 | XP_008758969.1 | 3, 4 | 499 | |||
XM_008760749.1 | XP_008758971.1 | 3, 4 | 484 | |||
XM_008760750.1 | XP_008758972.1 | 3, 4 | 498 | |||
GenBank mRNA | AY230250.1 | - | 2, 3 | 312 |
