siRNA Details
Gene Symbol: Bpifa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172031.1 | NP_742028.1 | 6 | 725 | ||
GenBank mRNA | AF393750.1 | - | 6 | 725 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bpifa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172031.1 | NP_742028.1 | 6 | 725 | ||
GenBank mRNA | AF393750.1 | - | 6 | 725 |