siRNA Details
Gene Symbol: Zc3hav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173045.2 | NP_766633.2 | 3 | 890 | ||
XM_006236295.2 | XP_006236357.1 | 3 | 951 | |||
GenBank mRNA | AF521008.1 | - | 3 | 574 | ||
BC131842.1 | - | 3 | 890 | 8372019 | ||
FQ230840.1 | - | 3 | 913 |
