siRNA Details
Gene Symbol: GPR85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146265.1 | NP_001139737.1 | 3 | 1139 | ||
NM_001146266.1 | NP_001139738.1 | 2 | 1029 | |||
NM_001146267.1 | NP_001139739.1 | 3 | 966 | |||
NM_018970.6 | NP_061843.3 | 3 | 1136 | |||
XM_005250451.2 | XP_005250508.1 | 2 | 1086 | |||
XM_017012352.1 | XP_016867841.1 | 3 | 1201 | |||
GenBank mRNA | AB040800.1 | - | 1 | 512 | ||
AF250237.2 | - | 3 | 1124 | |||
AK312587.1 | - | 2 | 904 | |||
AL161959.1 | - | 3 | 969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
