siRNA Details
Gene Symbol: Havcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173149.2 | NP_775172.1 | 3 | 1051 | ||
GenBank mRNA | AF035963.1 | - | 3 | 1052 | ||
BC061820.1 | - | 4 | 491 | 5599247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
