siRNA Details
Gene Symbol: Lama3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772008.1 | XP_008770230.1 | 59 | 8668 | ||
GenBank mRNA | U61261.1 | - | 21 | 3024 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
