siRNA Details
Gene Symbol: Grpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181440.1 | NP_852105.1 | 1, 2 | 106 | ||
GenBank mRNA | M17703.2 | - | 1, 2 | 106 | ||
M58653.1 | - | 1, 2 | 94 | |||
M58654.1 | - | 1, 2 | 106 |
Gene Symbol: Grpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203332.1 | NP_976077.1 | 106 | |||
XM_008763445.1 | XP_008761667.1 | 1, 2 | 102 | |||
GenBank mRNA | J02730.1 | - | 28 | |||
M31032.1 | - | 106 |
Gene Symbol: LOC100360165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237484.1 | XP_006237546.1 | 1, 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
