siRNA Details
Gene Symbol: Lppr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181634.2 | NP_853665.1 | 7 | 1695 | ||
GenBank mRNA | AY147866.1 | - | 7 | 1695 | ||
AY300027.1 | - | 7 | 1695 | |||
BC129083.1 | - | 5 | 1758 | |||
BC167750.1 | - | 7 | 1496 | 8366697 |
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170468.1 | NP_001163939.1 | 6 | 858 | ||
XM_006251958.2 | XP_006252020.1 | 6 | 827 | |||
XM_006251959.2 | XP_006252021.1 | 6 | 827 | |||
XM_006251960.2 | XP_006252022.1 | 5 | 707 | |||
XM_006251962.2 | XP_006252024.1 | 5 | 516 | |||
GenBank mRNA | FM033220.1 | - | 5 | 519 |
Gene Symbol: Cers3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127561.1 | NP_001121033.1 | 1081 | |||
GenBank mRNA | BC160913.1 | - | 1081 | 7457160 |
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013986.2 | NP_001014008.1 | 3 | 359 | ||
XM_006250482.2 | XP_006250544.1 | 3 | 382 | |||
GenBank mRNA | BC087114.1 | - | 3 | 313 | 7193786 | |
CB569988.1 | - | 371 | ||||
FQ217506.1 | - | 3 | 365 |
Gene Symbol: Ehmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212463.1 | NP_997628.1 | 7 | 1073 | ||
XM_006255954.2 | XP_006256016.2 | 7 | 1078 | |||
XM_006255955.2 | XP_006256017.2 | 7 | 1078 | |||
XM_006255956.2 | XP_006256018.2 | 7 | 1077 | |||
XM_006255957.1 | XP_006256019.1 | 8 | 902 | |||
XM_006255958.1 | XP_006256020.1 | 8 | 902 | |||
XM_006255959.1 | XP_006256021.1 | 8 | 902 |
Gene Symbol: Eif3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004283.1 | NP_001004283.1 | 14 | 1705 | ||
XM_006241938.2 | XP_006242000.1 | 14 | 1819 | |||
GenBank mRNA | BC079005.1 | - | 14 | 1705 | 7113045 |
Gene Symbol: Eif4e1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771512.1 | XP_008769734.1 | 2 | 63 |
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253139.2 | XP_006253201.1 | 9 | 2243 | ||
XM_008771256.1 | XP_008769478.1 | 9 | 2078 | |||
GenBank mRNA | BC166593.1 | - | 9 | 2023 | 7934031 |
Gene Symbol: LOC100361383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253741.2 | XP_006253803.1 | 4 | 887 | ||
XM_006253742.2 | XP_006253804.1 | 3 | 698 |
Gene Symbol: LOC100363112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177822.1 | NP_001171293.1 | 4 | 295 | ||
GenBank mRNA | AY325169.1 | - | 4 | 295 |
Gene Symbol: LOC100910978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253693.2 | XP_006253755.1 | 2251 | |||
XM_006253694.2 | XP_006253756.1 | 2062 | ||||
XM_008771523.1 | XP_008769745.1 | 756 |
Gene Symbol: LOC102547241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228757.2 | XP_006228819.1 | 7 | 3948 | ||
XM_006228758.2 | XP_006228820.1 | 8 | 4156 | |||
XM_006228759.2 | XP_006228821.1 | 8 | 4152 | |||
XM_008759232.1 | XP_008757454.1 | 7 | 3667 |
Gene Symbol: LOC499584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047948.1 | NP_001041413.1 | 2 | 397 | ||
GenBank mRNA | AY539953.1 | - | 2 | 397 |
Gene Symbol: LOC500956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025054.1 | NP_001020225.1 | 7 | 1410 | ||
XM_006242662.2 | XP_006242724.1 | 8 | 1115 | |||
XM_008765978.1 | XP_008764200.1 | 8 | 1154 | |||
GenBank mRNA | BC086569.1 | - | 7 | 1410 | 7300711 |
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106613.2 | NP_001100083.2 | 8 | 1136 | ||
XM_006236962.2 | XP_006237024.1 | 8 | 1210 | |||
GenBank mRNA | BC167050.1 | - | 8 | 1054 | 7114402 | |
EV776660.1 | - | 5 | 615 |
Gene Symbol: Prg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031619.2 | NP_113807.2 | 3 | 238 | ||
GenBank mRNA | CB703061.1 | - | 2 | 233 |
Gene Symbol: RGD1306556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249575.2 | XP_006249637.1 | 8 | 4402 | ||
XM_006249576.2 | XP_006249638.1 | 8 | 3139 |
Gene Symbol: Slc4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017049.1 | NP_058745.1 | 3 | 456 | ||
GenBank mRNA | J05167.1 | - | 3 | 456 |
Gene Symbol: Slc6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236903.2 | XP_006236965.1 | 14 | 5502 | ||
XM_008763123.1 | XP_008761345.1 | 16 | 5663 |
Gene Symbol: Styxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037788.1 | NP_001032877.1 | 4 | 323 | ||
XM_006249180.2 | XP_006249242.1 | 4 | 546 | |||
XM_006249181.2 | XP_006249243.1 | 4 | 508 | |||
XM_008769143.1 | XP_008767365.1 | 4 | 505 | |||
XM_008769144.1 | XP_008767366.1 | 2 | 196 | |||
XM_008769146.1 | XP_008767368.1 | 4 | 546 | |||
XM_008769147.1 | XP_008767369.1 | 2 | 360 | |||
GenBank mRNA | BC098678.1 | - | 4 | 323 | 7382696 |
Gene Symbol: Usp34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271196.1 | NP_001258125.1 | 15 | 1967 |
Gene Symbol: Zeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013164.1 | NP_037296.1 | 8 | 2883 | ||
GenBank mRNA | AY325179.1 | - | 8 | 2883 | ||
U51583.1 | - | 3130 |
Gene Symbol: Zfhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772567.1 | XP_008770789.1 | 1 | 6367 |
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255148.2 | XP_006255210.1 | 7 | 3948 | ||
XM_006255151.2 | XP_006255213.1 | 8 | 4152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
