siRNA Details
Gene Symbol: Hrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181369.2 | NP_852034.1 | 1 | 1834 | ||
GenBank mRNA | AY277279.1 | - | 1 | 1834 | ||
FQ216289.1 | - | 1 | 1928 | |||
FQ224148.1 | - | 1 | 1920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
