siRNA Details
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013060.3 | NP_037192.1 | 1 | 172 | ||
GenBank mRNA | AY321351.1 | - | 305 | |||
BC086391.1 | - | 167 | 7303450 | |||
D10863.1 | - | 1 | 172 | |||
FQ212969.1 | - | 190 | ||||
FQ213471.1 | - | 190 | ||||
FQ213662.1 | - | 190 | ||||
FQ220574.1 | - | 1 | 190 | |||
FQ227397.1 | - | 1 | 190 |

More Information
Other Species Targeted

Gene Ontology Categories:
