siRNA Details
Gene Symbol: Hba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013096.1 | NP_037228.1 | 2 | 164 | ||
GenBank mRNA | BC059150.1 | - | 2 | 163 | 6918464 | |
BC091567.1 | - | 151 | 7458326 | |||
BC160818.1 | - | 2 | 147 | 7375173 | ||
FQ209733.1 | - | 2 | 164 | |||
FQ209744.1 | - | 2 | 164 | |||
FQ209909.1 | - | 2 | 164 | |||
FQ209912.1 | - | 2 | 164 | |||
FQ210001.1 | - | 2 | 164 | |||
FQ210150.1 | - | 2 | 162 | |||
FQ210186.1 | - | 2 | 162 | |||
FQ210277.1 | - | 2 | 162 | |||
FQ210635.1 | - | 2 | 164 | |||
FQ210643.1 | - | 2 | 162 | |||
FQ210690.1 | - | 2 | 163 | |||
FQ210723.1 | - | 2 | 162 | |||
FQ210862.1 | - | 2 | 162 | |||
FQ210866.1 | - | 2 | 162 | |||
FQ211077.1 | - | 2 | 164 | |||
FQ211217.1 | - | 2 | 162 | |||
FQ211232.1 | - | 2 | 163 | |||
FQ211239.1 | - | 2 | 162 | |||
FQ211351.1 | - | 2 | 162 | |||
FQ211374.1 | - | 2 | 162 | |||
FQ211431.1 | - | 2 | 162 | |||
FQ211513.1 | - | 2 | 162 | |||
FQ211520.1 | - | 2 | 162 | |||
FQ211755.1 | - | 2 | 162 | |||
FQ211911.1 | - | 2 | 162 | |||
FQ211972.1 | - | 2 | 162 | |||
FQ212144.1 | - | 2 | 164 | |||
FQ212153.1 | - | 2 | 164 | |||
FQ212186.1 | - | 2 | 162 | |||
FQ212232.1 | - | 2 | 164 | |||
FQ212457.1 | - | 2 | 162 | |||
FQ212854.1 | - | 2 | 162 | |||
FQ213301.1 | - | 2 | 162 | |||
FQ214066.1 | - | 2 | 162 | |||
FQ214223.1 | - | 2 | 162 | |||
FQ216243.1 | - | 2 | 162 | |||
FQ217723.1 | - | 2 | 162 | |||
FQ217724.1 | - | 2 | 162 | |||
FQ217869.1 | - | 2 | 162 | |||
FQ217988.1 | - | 2 | 164 | |||
FQ218117.1 | - | 2 | 162 | |||
FQ218146.1 | - | 162 | ||||
FQ218257.1 | - | 2 | 218 | |||
FQ218612.1 | - | 2 | 161 | |||
FQ218968.1 | - | 2 | 164 | |||
FQ219715.1 | - | 2 | 162 | |||
FQ220786.1 | - | 2 | 162 | |||
FQ221656.1 | - | 2 | 163 | |||
FQ221912.1 | - | 2 | 164 | |||
FQ221952.1 | - | 2 | 162 | |||
FQ222431.1 | - | 2 | 162 | |||
FQ222454.1 | - | 2 | 162 | |||
FQ222672.1 | - | 2 | 162 | |||
FQ222750.1 | - | 2 | 164 | |||
FQ223017.1 | - | 2 | 163 | |||
FQ225427.1 | - | 2 | 163 | |||
FQ225429.1 | - | 2 | 164 | |||
FQ225440.1 | - | 2 | 163 | |||
FQ225452.1 | - | 2 | 162 | |||
FQ225777.1 | - | 2 | 162 | |||
FQ225808.1 | - | 2 | 165 | |||
FQ226118.1 | - | 2 | 164 | |||
FQ226173.1 | - | 2 | 164 | |||
FQ226175.1 | - | 2 | 162 | |||
FQ226203.1 | - | 2 | 162 | |||
FQ226615.1 | - | 2 | 162 | |||
FQ227082.1 | - | 2 | 162 | |||
FQ228301.1 | - | 2 | 162 | |||
FQ228436.1 | - | 162 | ||||
FQ230483.1 | - | 2 | 164 | |||
FQ230901.1 | - | 2 | 162 | |||
FQ231201.1 | - | 2 | 154 | |||
FQ231290.1 | - | 2 | 162 | |||
FQ232481.1 | - | 2 | 162 | |||
FQ234683.1 | - | 2 | 162 | |||
M17083.1 | - | 2 | 164 |
Gene Symbol: Hba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007722.1 | NP_001007723.1 | 164 | |||
GenBank mRNA | FQ209950.1 | - | 2 | 162 | ||
FQ210108.1 | - | 2 | 162 | |||
FQ210256.1 | - | 2 | 162 | |||
FQ210264.1 | - | 2 | 162 | |||
FQ210372.1 | - | 2 | 167 | |||
FQ210420.1 | - | 2 | 162 | |||
FQ210570.1 | - | 2 | 162 | |||
FQ210694.1 | - | 2 | 162 | |||
FQ211180.1 | - | 2 | 162 | |||
FQ211270.1 | - | 2 | 162 | |||
FQ211366.1 | - | 2 | 162 | |||
FQ211672.1 | - | 2 | 162 | |||
FQ211887.1 | - | 2 | 162 | |||
FQ213126.1 | - | 2 | 162 | |||
FQ214258.1 | - | 2 | 162 | |||
FQ215087.1 | - | 2 | 162 | |||
FQ217874.1 | - | 2 | 162 | |||
FQ218140.1 | - | 2 | 164 | |||
FQ218297.1 | - | 2 | 168 | |||
FQ218784.1 | - | 2 | 161 | |||
FQ221489.1 | - | 2 | 162 | |||
FQ222342.1 | - | 2 | 162 | |||
FQ222639.1 | - | 2 | 161 | |||
FQ222693.1 | - | 2 | 160 | |||
FQ223754.1 | - | 2 | 164 | |||
FQ223838.1 | - | 2 | 188 | |||
FQ224218.1 | - | 2 | 171 | |||
FQ224339.1 | - | 2 | 164 | |||
FQ224679.1 | - | 2 | 162 | |||
FQ225311.1 | - | 2 | 162 | |||
FQ225451.1 | - | 2 | 162 | |||
FQ225711.1 | - | 2 | 162 | |||
FQ225789.1 | - | 2 | 164 | |||
FQ226185.1 | - | 2 | 162 | |||
FQ226226.1 | - | 317 | ||||
FQ226238.1 | - | 2 | 152 | |||
FQ226556.1 | - | 2 | 162 | |||
FQ226574.1 | - | 2 | 162 | |||
FQ226576.1 | - | 2 | 162 | |||
FQ226594.1 | - | 2 | 162 | |||
FQ226619.1 | - | 2 | 162 | |||
FQ226919.1 | - | 2 | 160 | |||
FQ226927.1 | - | 2 | 162 | |||
FQ228017.1 | - | 2 | 164 | |||
FQ228292.1 | - | 2 | 162 | |||
FQ228298.1 | - | 2 | 162 | |||
FQ228306.1 | - | 2 | 162 | |||
FQ228588.1 | - | 2 | 162 | |||
FQ230499.1 | - | 2 | 162 | |||
FQ230501.1 | - | 2 | 162 | |||
FQ230536.1 | - | 2 | 162 | |||
FQ230819.1 | - | 2 | 164 | |||
FQ230868.1 | - | 2 | 162 | |||
FQ230877.1 | - | 2 | 164 | |||
FQ231222.1 | - | 2 | 162 | |||
FQ231239.1 | - | 2 | 157 | |||
FQ231246.1 | - | 2 | 162 | |||
FQ231250.1 | - | 2 | 162 | |||
FQ231268.1 | - | 2 | 161 | |||
FQ231572.1 | - | 2 | 162 | |||
FQ231902.1 | - | 2 | 162 | |||
FQ232213.1 | - | 2 | 162 | |||
FQ234805.1 | - | 2 | 162 | |||
FQ235117.1 | - | 2 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
