siRNA Details
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145771.2 | NP_665714.2 | 2 | 85 | ||
XM_008765674.1 | XP_008763896.1 | 2 | 269 | |||
GenBank mRNA | AF197128.1 | - | 2 | 79 | ||
AY738259.1 | - | 1 | 79 | |||
BC078840.1 | - | 2 | 85 | 7110224 | ||
FQ219187.1 | - | 1 | 74 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
