siRNA Details
Gene Symbol: Hist1h2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021839.1 | NP_068611.1 | 1 | 233 |
Gene Symbol: H2afx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109291.1 | NP_001102761.1 | 1 | 281 | ||
GenBank mRNA | CB713471.1 | - | 1 | 281 |

More Information
Other Species Targeted

Gene Ontology Categories:
