siRNA Details
Gene Symbol: Hba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013096.1 | NP_037228.1 | 2 | 224 | ||
GenBank mRNA | BC059150.1 | - | 2 | 223 | 6918464 | |
BC160818.1 | - | 2 | 207 | 7375173 | ||
FQ209733.1 | - | 2 | 224 | |||
FQ209744.1 | - | 2 | 224 | |||
FQ209909.1 | - | 2 | 224 | |||
FQ209912.1 | - | 2 | 224 | |||
FQ210001.1 | - | 2 | 224 | |||
FQ210150.1 | - | 2 | 222 | |||
FQ210186.1 | - | 2 | 222 | |||
FQ210277.1 | - | 2 | 222 | |||
FQ210349.1 | - | 2 | 222 | |||
FQ210635.1 | - | 2 | 224 | |||
FQ210643.1 | - | 2 | 222 | |||
FQ210690.1 | - | 2 | 223 | |||
FQ210723.1 | - | 2 | 222 | |||
FQ210862.1 | - | 2 | 222 | |||
FQ210866.1 | - | 2 | 222 | |||
FQ211077.1 | - | 2 | 224 | |||
FQ211217.1 | - | 2 | 222 | |||
FQ211220.1 | - | 2 | 224 | |||
FQ211232.1 | - | 2 | 223 | |||
FQ211239.1 | - | 2 | 222 | |||
FQ211351.1 | - | 2 | 222 | |||
FQ211374.1 | - | 2 | 222 | |||
FQ211431.1 | - | 2 | 222 | |||
FQ211513.1 | - | 2 | 222 | |||
FQ211755.1 | - | 2 | 222 | |||
FQ211911.1 | - | 2 | 222 | |||
FQ211972.1 | - | 2 | 222 | |||
FQ212025.1 | - | 2 | 224 | |||
FQ212144.1 | - | 2 | 224 | |||
FQ212153.1 | - | 2 | 224 | |||
FQ212186.1 | - | 2 | 222 | |||
FQ212232.1 | - | 2 | 224 | |||
FQ212457.1 | - | 2 | 222 | |||
FQ212854.1 | - | 2 | 222 | |||
FQ213301.1 | - | 2 | 222 | |||
FQ214066.1 | - | 2 | 222 | |||
FQ214223.1 | - | 2 | 222 | |||
FQ216243.1 | - | 2 | 222 | |||
FQ217723.1 | - | 2 | 222 | |||
FQ217724.1 | - | 2 | 221 | |||
FQ217869.1 | - | 2 | 222 | |||
FQ218117.1 | - | 2 | 222 | |||
FQ218257.1 | - | 2 | 278 | |||
FQ218612.1 | - | 2 | 221 | |||
FQ218968.1 | - | 2 | 224 | |||
FQ219715.1 | - | 2 | 222 | |||
FQ221656.1 | - | 2 | 223 | |||
FQ221912.1 | - | 2 | 224 | |||
FQ221952.1 | - | 2 | 222 | |||
FQ222431.1 | - | 2 | 222 | |||
FQ222454.1 | - | 2 | 222 | |||
FQ222672.1 | - | 2 | 222 | |||
FQ222750.1 | - | 2 | 224 | |||
FQ223017.1 | - | 2 | 223 | |||
FQ224581.1 | - | 2 | 222 | |||
FQ225427.1 | - | 2 | 223 | |||
FQ225429.1 | - | 2 | 224 | |||
FQ225440.1 | - | 2 | 223 | |||
FQ225452.1 | - | 2 | 222 | |||
FQ225777.1 | - | 2 | 222 | |||
FQ225808.1 | - | 2 | 225 | |||
FQ226118.1 | - | 2 | 224 | |||
FQ226173.1 | - | 2 | 224 | |||
FQ226175.1 | - | 2 | 222 | |||
FQ226203.1 | - | 2 | 222 | |||
FQ226615.1 | - | 2 | 222 | |||
FQ227082.1 | - | 2 | 222 | |||
FQ228301.1 | - | 2 | 222 | |||
FQ228436.1 | - | 222 | ||||
FQ230483.1 | - | 2 | 224 | |||
FQ230901.1 | - | 2 | 222 | |||
FQ231201.1 | - | 2 | 214 | |||
FQ231290.1 | - | 2 | 222 | |||
FQ232272.1 | - | 2 | 222 | |||
FQ232481.1 | - | 2 | 222 | |||
FQ234683.1 | - | 2 | 222 | |||
M17083.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
