siRNA Details
Gene Symbol: Cxcl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033882.1 | NP_001029054.1 | 3 | 283 | ||
NM_001033883.1 | NP_001029055.1 | 3 | 283 | |||
NM_022177.3 | NP_071513.1 | 3 | 283 | |||
GenBank mRNA | AF189724.1 | - | 3 | 204 | ||
AF209976.1 | - | 3 | 234 | |||
AF217564.1 | - | 3 | 255 | |||
BC078737.1 | - | 3 | 282 | 7128141 | ||
CK475409.1 | - | 3 | 283 | |||
CV081127.1 | - | 3 | 250 |
