siRNA Details
Gene Symbol: Cklf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139111.1 | NP_620811.1 | 2 | 331 | ||
XM_008772240.1 | XP_008770462.1 | 2 | 320 | |||
GenBank mRNA | AF253065.2 | - | 2 | 331 |
Gene Symbol: Cmtm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013142.1 | NP_001013160.1 | 5 | 692 | ||
GenBank mRNA | BC078975.1 | - | 5 | 692 | 7111936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
