siRNA Details
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107324.2 | NP_001100794.2 | 29 | 6256 | ||
XM_006253446.2 | XP_006253508.1 | 30 | 6315 | |||
GenBank mRNA | AY064511.1 | - | 1 | 154 | ||
BC097278.1 | - | 1 | 808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
