siRNA Details
Gene Symbol: LOC259245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147213.1 | NP_671746.1 | 3 | 293 | ||
GenBank mRNA | AB039826.1 | - | 3 | 293 |
Gene Symbol: Mup5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203325.1 | NP_976070.1 | 293 | |||
GenBank mRNA | AB039827.1 | - | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
