siRNA Details
Gene Symbol: Ugt2b17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153314.2 | NP_695226.2 | 284 | |||
XM_006250800.2 | XP_006250862.1 | 1 | 1378 | |||
XM_008769998.1 | XP_008768220.1 | 1 | 1384 | |||
GenBank mRNA | M31109.1 | - | 284 | |||
Y00156.1 | - | 1 | 328 |
Gene Symbol: Ugt2b37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770015.1 | XP_008768237.1 | 1 | 317 | ||
GenBank mRNA | M33746.1 | - | 1 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
