siRNA Details
Gene Symbol: Cstb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012838.2 | NP_036970.1 | 2 | 177 | ||
GenBank mRNA | BC084725.1 | - | 2 | 165 | 7318993 | |
CF976177.1 | - | 2 | 178 | |||
FQ221069.1 | - | 2 | 137 | |||
FQ222613.1 | - | 2 | 132 | |||
X54737.1 | - | 2 | 169 |
