siRNA Details
Gene Symbol: Fgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012846.1 | NP_036978.1 | 3 | 468 | ||
XM_006254650.2 | XP_006254712.1 | 4 | 772 | |||
XM_006254651.2 | XP_006254713.1 | 3 | 1589 | |||
XM_006254652.2 | XP_006254714.1 | 3 | 789 | |||
XM_006254653.2 | XP_006254715.1 | 3 | 624 | |||
XM_006254654.2 | XP_006254716.1 | 4 | 776 | |||
GenBank mRNA | X14232.1 | - | 3 | 468 |
