siRNA Details
Gene Symbol: Bglap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013414.1 | NP_038200.1 | 2 | 134 | ||
XM_006232594.2 | XP_006232656.1 | 3 | 3191 | |||
GenBank mRNA | M11777.1 | - | 2 | 114 | ||
X04141.1 | - | 2 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
