siRNA Details
Gene Symbol: Psmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017285.1 | NP_058981.1 | 2 | 205 | ||
GenBank mRNA | BC084723.1 | - | 1 | 128 | 7319054 | |
D21800.1 | - | 2 | 205 | |||
FQ211229.1 | - | 2 | 162 | |||
FQ232780.1 | - | 2 | 201 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011971, BC014783, AK010488, AK153162, BC098233, and others.
Gene Ontology Categories:
