siRNA Details
Gene Symbol: Olr414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020106.1 | NP_064491.1 | 1 | 74 |
Gene Symbol: Olr416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000387.1 | NP_001000387.1 | 1 | 74 |

More Information
Other Species Targeted

Gene Ontology Categories:
