siRNA Details
Gene Symbol: Insig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022392.1 | NP_071787.1 | 1 | 264 | ||
GenBank mRNA | BC078827.1 | - | 1 | 274 | 7125208 | |
L13619.1 | - | 1 | 264 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Insig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022392.1 | NP_071787.1 | 1 | 264 | ||
GenBank mRNA | BC078827.1 | - | 1 | 274 | 7125208 | |
L13619.1 | - | 1 | 264 |