siRNA Details
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030996.1 | NP_112258.1 | 1 | 145 | ||
GenBank mRNA | AF004218.1 | - | 1 | 145 | ||
AF067769.1 | - | 1 | 197 | |||
AF087827.1 | - | 1 | 197 | |||
BC061978.1 | - | 1 | 134 | 5623693 |
