siRNA Details
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031598.3 | NP_113786.3 | 3 | 140 | ||
XM_006239146.2 | XP_006239208.1 | 4 | 306 | |||
GenBank mRNA | AY651027.1 | - | 2 | 84 | ||
D00523.1 | - | 3 | 124 | |||
FQ220554.1 | - | 2 | 356 | |||
FQ227448.1 | - | 3 | 140 | |||
FQ234749.1 | - | 4 | 238 | |||
M25148.1 | - | 3 | 140 | |||
X74364.1 | - | 1 | 20 |
