siRNA Details
Gene Symbol: Kcng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133426.2 | NP_596917.2 | 1, 2 | 698 | ||
GenBank mRNA | AB070605.1 | - | 1, 2 | 392 | ||
AF454550.1 | - | 1, 2 | 882 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
