siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 2 | 718 | ||
NM_001270590.1 | NP_001257519.1 | 2 | 403 | |||
NM_001270591.1 | NP_001257520.1 | 3 | 181 | |||
NM_001270592.1 | NP_001257521.1 | 3 | 178 | |||
NM_001270593.1 | NP_001257522.1 | 2 | 115 | |||
NM_001270594.1 | NP_001257523.1 | 2 | 112 | |||
NM_134390.3 | NP_599217.1 | 2 | 721 | |||
XM_006236455.2 | XP_006236517.1 | 2 | 104 | |||
GenBank mRNA | AF370882.1 | - | 1 | 74 | ||
BC059116.1 | - | 2 | 220 | 6920949 | ||
FM070640.1 | - | 2 | 102 | |||
FM086181.1 | - | 3 | 165 | |||
FN804490.1 | - | 3 | 164 | |||
FQ212987.1 | - | 2 | 110 | |||
FQ219149.1 | - | 3 | 165 | |||
FQ219532.1 | - | 3 | 165 | |||
FQ219576.1 | - | 2 | 109 | |||
FQ220000.1 | - | 2 | 294 | |||
FQ233370.1 | - | 3 | 175 | |||
FQ235102.1 | - | 2 | 127 |
