siRNA Details
Gene Symbol: Vom1r100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173296.1 | NP_775418.1 | 3 | 277 | ||
GenBank mRNA | AY510294.1 | - | 1 | 142 | ||
U36895.1 | - | 3 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
