siRNA Details
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007459.3 | NP_031485.3 | 2, 3 | 324 | ||
GenBank mRNA | AK009735.1 | - | 2, 3 | 336 | ||
AK088500.1 | - | 2, 3 | 258 | |||
AK128972.1 | - | 2, 3 | 267 | |||
AK152463.1 | - | 2, 3 | 257 | |||
AK170980.1 | - | 2, 3 | 271 | |||
AK173054.1 | - | 2, 3 | 261 | |||
BY729047.1 | - | 2, 3 | 324 | |||
X14972.1 | - | 2, 3 | 294 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031008, XM_008760010, BC081786, X53773.
Gene Ontology Categories:
