siRNA Details
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007459.3 | NP_031485.3 | 3 | 332 | ||
GenBank mRNA | AK009735.1 | - | 3 | 344 | ||
AK088500.1 | - | 3 | 266 | |||
AK128972.1 | - | 3 | 275 | |||
AK152463.1 | - | 3 | 265 | |||
AK170980.1 | - | 3 | 279 | |||
AK173054.1 | - | 3 | 269 | |||
BY729047.1 | - | 3 | 332 | |||
X14972.1 | - | 3 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
