siRNA Details
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007517.4 | NP_031543.2 | 2, 3 | 458 | ||
GenBank mRNA | AK144497.1 | - | 1 | 410 | ||
AK154707.1 | - | 2, 3 | 327 | |||
AK158035.1 | - | 2, 3 | 249 | |||
AK167335.1 | - | 2, 3 | 248 | |||
AK171011.1 | - | 2, 3 | 261 | |||
AK171184.1 | - | 2, 3 | 253 | |||
AK171202.1 | - | 2, 3 | 267 | |||
AK171249.1 | - | 2, 3 | 247 | |||
AK171541.1 | - | 2, 3 | 248 | |||
AK171621.1 | - | 2, 3 | 239 | |||
BC016485.1 | - | 2, 3 | 251 | 3962982 | ||
U41736.1 | - | 2, 3 | 256 |

More Information
Other Species Targeted

Gene Ontology Categories:
